Learn More
AdipoGen (R,R)-Hymeglusin (0.5 mg)
SDP
Supplier:  AdipoGen AGCN20103MC05
Encompass_Preferred
Chemical. CAS: 29066-42-0. Formula: C18H28O5. MW: 324.4. Isolated from fungal strain Scopulariopsis sp. F-244. beta-Lactone antibiotic. Antibacterial and antifungal. Specific and irreversible inhibitor of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase in vitro and in vivo by covalently modifying the active Cys129 residue of the enzyme. Mevalonate biosynthesis inhibitor.
Catalog No. 50-205-3461