Learn More
Cayman Chemical R S-AMPAhydrate 10mg
SDP
Supplier:  Cayman Chemical 1457110 MG
Encompass_Preferred
The defining agonist for the AMPA subgroup of ionotropic glutamate receptors, which mediate excitatory neurotransmission; contains an equal mixture of the active (S)-enantiomer and the inactive (R)-enantiomer
Catalog No. 50-297-2975