Learn More
Medchemexpress LLC HY-126047A 10mg Medchemexpress, (R,S)-Anatabine CAS:2743-90-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY126047A10MG
Encompass_Preferred
Medchemexpress, HY-126047A 10mg (R,S)-Anatabine CAS:2743-90-0 (R,S)-Anatabine is a a minor tobacco alkaloid found in the Solanaceae family of plants that can be used as a specific marker for the detection of tobacco use. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9715