Learn More
Medchemexpress LLC R S-Anatabine-d4 | 1020719-11-2 | 1MG
SDP
Supplier:  Medchemexpress LLC HY126047AS1MG
Encompass_Preferred
(R S)-Anatabine-d4 is a deuterated labeled (R S)-Anatabine (HY-126047A) (R S)-Anatabine is a a minor tobacco alkaloid found in the Solanaceae family of plants that can be used as a specific marker for the detection of tobacco use[1][2]
Catalog No. 50-002-35923