Learn More
Enzo Life Sciences RS-Sulforaphane, (25mg), CAS Number: 4478-93-7 (unspecified stereo)
SDP
Supplier:  Enzo Life Sciences ALX350232M025
Encompass_Preferred
Carcinogenesis inhibitor. R,S-Sulforaphane is a synthetic version of the naturally occurring compound, R-sulforaphane, from Broccoli. It is a racemic mixture of both the R and S isomers.Potent, selective inducer of phase II detoxification enzymes. Inhibits chemically induced mammary tumor formation in rats. Inhibits LPS-induced HMGB1 (high mobility group box 1) secretion. Alternative Name: DL-Sulforaphane, 1-Isothiocyanato-4-methylsulfinyl-butane. Formula: C6H11NOS2. MW: 177.3. CAS Number: 4478-93-7 (unspecified stereo). MI: 14: 8962. Purity: ≥98% (UPLC). Appearance: Slightly yellowish liquid. Solubility: Soluble in methanol or DMSO. Long Term Storage: -20°C
Catalog No. NC0198966