Learn More
Medchemexpress LLC (R)-(+)-tert-Butylsulfinamide | 196929-78-9 | 100.0% | 121.20 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY40228
Encompass_Preferred
(R)-(+)-tert-Butylsulfinamide is a chiral reagent commonly used in the asymmetric synthesis of compounds such as amines, amino acids, and amino alcohols. It achieves stereoselective reactions by condensing with carbonyl groups to form sulfinyl imine intermediates, which can then be removed under acidic conditions to obtain the target product.
  • Chiral reagent for asymmetric synthesis.
  • Used in the synthesis of amines, amino acids, and amino alcohols.
  • Achieves stereoselective reactions.
  • Participates in cycloaddition and cyclization reactions of enynes and diynes through asymmetric induction, N-alkylation, and deprotection or oxidation under mild conditions to construct polycyclic scaffold libraries related to alkaloid and terpenoid natural products.
Catalog No. 50-004-03097