Learn More
Medchemexpress LLC (R)-thalidomide | 2614-06-4 | 99.8% | 258.23 g/mol | C13H10N2O4 | 5 MG

Supplier: Medchemexpress LLC HY14658B5MG
(R)-Thalidomide is the R-enantiomer of thalidomide supplied as a high-purity research reagent. It is described as having psychomotor-stabilizing properties and is provided as a white to off-white solid for biochemical and pharmacological studies (research use only).
- R-enantiomer of thalidomide.
- High purity (99.8%).
- White to off-white solid appearance.
- Molecular formula C13H10N2O4; molecular weight 258.23 g/mol.
- Provided for biochemical and pharmacological research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.