Learn More
Cayman Chemical R-TrItyl glycIdyl ether 1g
SDP
Supplier:  Cayman Chemical 341311 G
Encompass_Preferred
A synthetic precursor; has been used in the synthesis of glycerophospholipids, as well as compounds with antiviral and antimalarial activities
Catalog No. 50-294-6571