Learn More
Medchemexpress LLC (R)-UT-155 | 2031161-54-1 | 98.1% | 405.35 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY112895A50MG
Encompass_Preferred
(R)-UT-155 is a selective androgen receptor degrader (SARD) ligand. It is less active than its S-isomer. This product is for research use only and is not sold to patients.
  • Selective androgen receptor degrader (SARD) ligand.
  • Antagonist activity at human androgen receptor expressed in HEK293 cells with an IC50 of 0.598 μM, assessed as inhibition of R1881-induced receptor transactivation after 24 hours by luciferase reporter gene assay.
Catalog No. 50-004-39215