Learn More
Medchemexpress LLC (R)-V-0219 hydrochloride | C20H26ClF3N4O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY143312D100MG
Encompass_Preferred
(R)-V-0219 hydrochloride is an enantiomer of V-0219, functioning as an orally active and positive allosteric modulator (PAM) of the GLP Receptor-1 (GLP-1R). It is noted for activating calcium fluxes in HEK cells stably expressing hGLP-1R.
  • Induces receptor activation stimulated by GLP-1 with an EC50 value of 10 nM.
  • Activates calcium fluxes in HEK cells stably expressing hGLP-1R.
Catalog No. 50-003-54083