Learn More
Medchemexpress LLC (R)-vanzacaftor | 2374124-48-6 | 98.1% | 617.76 | C32H39N7O4S | 5 MG

Supplier: Medchemexpress LLC HY145603A5MG
(R)-Vanzacaftor is a research-grade small-molecule regulator of the cystic fibrosis transmembrane conductance regulator (CFTR). Supplied as a solid with defined purity and physicochemical properties, it is intended for biochemical and cell-based assays. For research use only.
- Regulates cystic fibrosis transmembrane conductance regulator (CFTR).
- High purity: 98.1%.
- Molecular formula: C32H39N7O4S; molecular weight: 617.76 g·mol⁻¹.
- Solubility in DMSO: 100 mg/mL; ultrasonic agitation recommended.
- Appearance: off-white to light yellow solid.
- Storage: store under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.