Learn More
Medchemexpress LLC (R)-viloxazine-d5 hydrochloride | 1246815-04-2 | 95.0% | 278.79 | C13H15D5ClNO3 | 1MG
SDP
Supplier:  Medchemexpress LLC HY125784AS1MG
Encompass_Preferred
(R)-Viloxazine-d5 hydrochloride is the deuterium-labeled R-enantiomer of viloxazine hydrochloride supplied as the hydrochloride salt for use as an isotopic tracer or internal standard in quantitative analysis. It is intended for research use and is commonly applied in NMR, GC-MS, and LC-MS method development. The compound has CAS 1246815-04-2, formula C13H15D5ClNO3, and molecular weight 278.79.
  • Deuterium-labeled with five deuteriums for tracer and quantitative applications.
  • R-enantiomer suitable for stereospecific studies.
  • Hydrochloride salt form for improved stability and handling.
  • Intended as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • Reported purity 95.0% as listed by distributor sources.
  • Provided in small analytical quantities for standards and method development.
Catalog No. 50-002-35137