Learn More
Medchemexpress LLC (R)-vu 6008667 | 2097818-14-7 | 99.8% | 438.85 | C24H17ClF2N2O2 | 100 MG

Supplier: Medchemexpress LLC HY101281B100MG
This research compound is the (R)-enantiomer of an mAChR M5 negative allosteric modulator. It is reported to be devoid of M5 NAM activity (IC50 > 10 μM) and is supplied as a high-purity solid for use in in vitro and in vivo studies.
- Less active R-enantiomer; no M5 NAM activity (IC50 > 10 μM).
- Molecular weight 438.85.
- Formula C24H17ClF2N2O2.
- Purity 99.79%.
- White to off-white solid appearance.
- Soluble in DMSO at 100 mg/mL; ultrasonication recommended.
- Storage: solid at 4°C under nitrogen; stock solutions -80°C (6 months) or -20°C (1 month).
- Available in small pack sizes (5-100 mg) suitable for research workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.