Learn More
Medchemexpress LLC (R)-VU 6008667 | 2097818-14-7 | 99.8% | 438.85 | C24H17ClF2N2O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY101281B25MG
Encompass_Preferred
(R)-VU 6008667 is the R-enantiomer of VU 6008667 used for research applications. It is reported as the less active stereoisomer and is devoid of M5 negative allosteric modulator (NAM) activity (IC50 > 10 μM). The compound is provided with high purity and is intended for in vitro pharmacology and neuroscience studies.
  • High analytical purity suitable for research applications.
  • Characterized by molecular weight 438.85 g/mol.
  • Limited M5 NAM activity (IC50 > 10 μM), useful as a control compound.
  • Supplied in small quantities for assay screening and medicinal chemistry.
  • Soluble in DMSO for preparation of concentrated stock solutions.
Catalog No. 50-002-12565