Learn More
Medchemexpress LLC (R)-(+)-warfarin-d5 | 773005-79-1 | MFCD08063545 | 99.2% | 313.36 | C19H11D5O4 | 1 MG

Supplier: Medchemexpress LLC HYW015998S1MG
(R)-(+)-Warfarin-d5 is a deuterium-labeled R-enantiomer of warfarin supplied as a solid reference standard for analytical and quantitative applications. It is intended for use as a tracer or internal standard in NMR, GC-MS, and LC-MS workflows.
- High isotopic labeling and analytical purity (99.2%).
- Suitable as a tracer and internal standard for NMR, GC-MS, and LC-MS.
- Solid, white to off-white powder for easy handling.
- Molecular weight 313.36 and formula C19H11D5O4 for unambiguous identification.
- Storage: powder -20°C (3 years); in solvent -80°C (6 months) or -20°C (1 month).
- Controlled substance restrictions may apply in some territories.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.