Learn More
Medchemexpress LLC (R)-WM-586 | C20H20F3N5O3S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY153728A1MG
Encompass_Preferred
(R)-WM-586 is the R-enantiomer of WM-586, functioning as a covalent WDR5 inhibitor. This compound is primarily utilized in research for various cancers, including neuroblastoma, breast cancer, bladder cancer, and colorectal cancer, by specifically decreasing the interaction between WDR5 and MYC.
  • Functions as a covalent WDR5 inhibitor
  • Specifically decreases WDR5 and MYC interaction with an IC50 of 101 nM
  • Used in research for various cancers
  • Does not affect the binding of WDR5 to RBBP5
Catalog No. 50-003-76020