Learn More
TARGETMOL CHEMICALS INC R-ZANUBRUTINIB 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T134475MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (R)-Zanubrutinib ((R)-BGB-3111) is a selective inhibitor of Bruton tyrosine kinase (BTK). purity: 98%
Catalog No. 50-258-1347