Learn More
TARGETMOL CHEMICALS INC R-ZINC-3573 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2922125MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (R)-ZINC-3573 is a potent and highly selective MRGPRX2 probe that activates endogenous MRGPRX2 in a human mast cell line, inducing degranulation and calcium release. Purity 100%
Catalog No. 50-287-9440