Learn More
Cayman Chemical R-ZINC3573 10mg
SDP
Supplier:  Cayman Chemical 4131210 MG
Encompass_Preferred
An MRGPRX2 agonist; induces β-arrestin recruitment in a reporter assay using MRGPRX2-expressing HTLA cells and calcium mobilization in LAD 2 mast cells (EC50s = 0.74 and 1 µM, respectively); selective for MRGPRX2 over 21 other GPCRs; induces degranulation of LAD 2 mast cells
Catalog No. 50-294-5936