Learn More
Medchemexpress LLC HY-103096 5mg Medchemexpress, R162 CAS:64302-87-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1030965MG
Encompass_Preferred
Medchemexpress, HY-103096 5mg R162 CAS:64302-87-0 R162 is a potent inhibitor of glutamate dehydrogenase 1 (GDH1/GLUD1), with anti-cancer properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8033