Learn More
Alkali Scientific R2A Agar, 1 Kg
SDP
Supplier:  Alkali Scientific RAA001CP
Encompass
R2A Agar (Low Nutrient Media) is a specialized microbiological growth medium used for the recovery and enumeration of heterotrophic bacteria, particularly from treated potable water and pharmaceutical process water. Its low-nutrient composition prevents the rapid overgrowth of bacteria, allowing slow-growing or stressed microorganisms to recover. The 1 kg package provides sufficient volume for large-scale studies, enabling the cultivation of slow-growing or oligotrophic organisms. The agar is used in routine water quality testing, environmental monitoring, and heterotrophic plate count (HPC) procedures to detect and enumerate bacteria in water samples.
Catalog No. 50-006-61882