Learn More
Selleck Chemical LLC R406 10mM/1mL 841290-81-1
SDP
Supplier:  Selleck Chemical LLC S219410MM/1ML
Encompass_Preferred
R406 is a potent Syk inhibitor with IC50 of 41 nM in cell-free assays, strongly inhibits Syk but not Lyn, 5-fold less potent to Flt3. R406 induces apoptosis. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2151