Learn More
Apexbio Technology LLC R406, 2mg. Cas: 841290-81-1 MFCD: MFCD18385012. SYK inhibitor,potent and ATP-competitive
SDP
Supplier:  Apexbio Technology LLC A85462
Encompass_Preferred
R406is a potent SYK inhibitorSpleen tyrosine kinase (SYK) is a non-receptor tyrosine kinase mainly expressed in hematopoietic cells. It transmits signals from a variety of cell surface receptors including CD74, Fc receptor and integrins. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5160