Learn More
Apexbio Technology LLC R406 (free base),5mg CAS# 841290-80-0
SDP
Supplier:  Apexbio Technology LLC A58805
Encompass_Preferred
R406 is a potent SYK inhibitorSpleen tyrosine kinase (SYK) is a non-receptor tyrosine kinase mainly expressed in hematopoietic cells. It transmits signals from a variety of cell surface receptors including CD74, Fc receptor and integrins. It is crucial for adaptive immune response, and also very important for cellular adhesion, innate immune recognition, osteoclast maturation, platelet activation and vascular development. [1] Functional abnormality of SYK has been implicated in several blood malignancies. Constitutively active SYK can transform B cells. SYK inhibition can be beneficial for patients with blood cancers and autoimmune diseases. R406 is a potent inhibitor of IgE and IgG mediated Fc receptor activation with EC50 for degranulation of 56-64 nM. [2] Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0488