Learn More
Selleck Chemical LLC R406 (free base) 50mg 841290-80-0
SDP
Supplier:  Selleck Chemical LLC S153350MG
Encompass_Preferred
R406 (free base) is a potent Syk inhibitor with IC50 of 41 nM in a cell-free assay, strongly inhibits Syk but not Lyn, 5-fold less potent to Flt3. R406 (free base) triggers apoptosis. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-791-7