Learn More
Apexbio Technology LLC R428, 10mg. Cas: 1037624-75-1 MFCD: MFCD21608463. Selective Axl inhibitor
SDP
Supplier:  Apexbio Technology LLC A832910
Encompass_Preferred
R428 is a selective Axl inhibitor with an IC50 of 14 nM, more than 50-fold sensitivity for Axl than Abl, Mer, Tyro3, InsR, EGFR, HER2, and PDGFR. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5112