Learn More
Apexbio Technology LLC R428, 1037624-75-1, MFCD21608463, 5 mg
SDP
Supplier:  Apexbio Technology LLC A83295
Encompass_Preferred
MF: C30H34N8, MW: 506.64, Solubility: 25mg/mL in DMSO with gentle warming. R428 is a selective Axl inhibitor with an IC50 of 14 nM, more than 50-fold sensitivity for Axl than Abl, Mer, Tyro3, InsR, EGFR, HER2, and PDGFR.
Catalog No. 50-101-4610