Learn More
Selleck Chemical LLC Bemcentinib (R428) 1mg 1037624-75-1 BGB324
SDP
Supplier:  Selleck Chemical LLC S28411MG
Encompass_Preferred
Bemcentinib (R428, BGB324) is an inhibitor of Axl with IC50 of 14 nM, >100-fold selective for Axl versus Abl. Selectivty for Axl is also greater than Mer and Tyro3 (50-to-100- fold more selective) and InsR, EGFR, HER2, and PDGFR? (100- fold more selective). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0532629