Learn More
Lauda Brinkmann Lp RA 12 REFRIGERATED CIRCULATOR
Supplier:  Lauda Brinkmann Lp L000651
Encompass_Preferred
Refrigerated Circulator, Alpha Series, Model RA 12, Digital control interface with simple 3 button operation, 12 L bath size, 325 Watts of cooling capacity at 20oC, 1.15 kW Heater, Temperature range of -25 to 85 deg.C, Flow rate from 5 to 15 L/min., 115V; 6 This item is non returnable*
Catalog No. 50-931-212
Only null left
Add to Cart
Request a Quote