Learn More
Selleck Chemical LLC RA-190
SDP
Supplier:  Selleck Chemical LLC S685125MG
Encompass_Preferred
RA190 a bis-benzylidine piperidon is a potent selective and oral effective inhibitor of proteasome ubiquitin receptor RPN13/ADRM1 with anticancer activity RA190 triggers ER stress response p53/p21 signaling axis and autophagy in multiple myeloma cells
Catalog No. 50-000-02604