Learn More
ATCC rAAV2 RSS; Reference material
Supplier:  ATCC VR1616
Encompass
Recombinant Adeno-associated virus 2 was developed by the Adeno-associated Virus Reference Standard Working Group (AAVRSWG) to use as reference material in gene therapy research. The supply of this product is limited to 3 vials per customer per year. This limit will be re-evaluated when supply is replenished. We apologize for any inconvenience this causes.
Catalog No. 50-238-2702