Learn More
Medchemexpress LLC Rab1b protein, human (HEK293, Fc) | 97.9% | 50 UG
SDP
Supplier:  Medchemexpress LLC HYP7599350UG
Encompass_Preferred
Recombinant human RAB1B protein produced in HEK293 mammalian cells and fused to an Fc tag. Supplied for research applications studying vesicular trafficking, protein-protein interactions, and cell signaling, the material is characterized for molecular weight and purity and provided in laboratory-scale quantities.
  • Recombinant human RAB1B expressed in HEK293 mammalian cells.
  • Fc tag facilitates detection and affinity purification.
  • Characterized molecular weight of approximately 58 kDa.
  • High purity suitable for biochemical and interaction studies.
  • Available in small lab-scale quantities such as 50 μg.
  • Quality-assessed by reducing SDS-PAGE for batch verification.
Catalog No. 50-002-92475