Learn More
Immunoreagents Inc RABBIT A-LLAMA IGG AP
Supplier:  Immunoreagents Inc RBXLL003D
Encompass_Preferred
Secondary Antibody; Rabbit anti-llama IgG (H&L) - Affinity Pure, Host: Rabbit, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-271-294