Learn More
ACROBiosystems Rabbit Angiopoietin-2 / ANGPT2 Protein, His Tag (MALS verified)
Supplier: ACROBiosystems AN2R52H4100UG
Rabbit Angiopoietin-2 Protein, His Tag (AN2-R52H4) is expressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession # G1T9B3-1).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.