Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Camelid VHH Antibody Plate (Clear, 8×12 strip)
Supplier:  Genscript Corporation L00946
Encompass
The product is used for rapid capture of VHHs in different samples. Camelid VHH Antibody (also called single-domain antibody (sdAb), or Nanobody by Ablynx) is a peptide chain about 110 amino acids in length, containing one variable domain of a heavy-chain antibody (a VHH fragment). Like a whole antibody, the camelid VHH antibody is able to bind selectively to a specific antigen. Due to its small size, simple production and high affinity, the camelid VHH antibody has a broad range of applications in biotechnical and therapeutic research fields.
Catalog No. NC2229429