Learn More
Immunology Consultants anti-Human Ferritin Antibody (Rabbit) - Affinity Purified
SDP
Supplier:  Immunology Consultants RF80A
Encompass_Preferred
RF‑80A is an affinity‑purified rabbit polyclonal antibody specific for human ferritin, a key intracellular iron‑storage protein and an important biomarker in clinical and research applications. The purification process enriches high‑affinity immunoglobulins, ensuring strong specificity, reduced background, and consistent performance across immunoassay platforms. This antibody is well suited for ELISA, Western blotting, immunohistochemistry, and other ferritin‑detection workflows requiring reliable and sensitive recognition. Produced by Immunology Consultants and supplied in a 1 mg format, RF‑80A provides a high‑quality reagent for laboratories studying iron metabolism, inflammation, or ferritin‑related physiological and pathological processes
Catalog No. 50-224-8823