Learn More
Immunology Consultants anti-Human IgG h+l Antibody (Rabbit) - Affinity Purified
SDP
Supplier:  Immunology Consultants RGHL80A
Encompass_Preferred
RGHL‑80A is an affinity‑purified rabbit antibody targeting human IgG heavy and light chains, suitable for use as a secondary antibody in immunoassays. This reagent provides strong specificity, low background, and consistent performance in ELISA, Western blotting, and immunodetection workflows involving human primary antibodies. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑80A is a reliable reagent for human IgG detection.
Catalog No. 50-224-8878