Learn More
Alpha Diagnostic International Rabbit Anti-rat AQP1 IgG #1 (aff pure) | 100 uL
SDP
Supplier:  Alpha Diagnostic International AQP11A
Encompass
19-aa peptide of Rat AQP1 (protein accession #P29975, refs 1) Designated (AQP11-P or or control peptide) conjugated to KLH; epitope location ~ C-terminus.
Catalog No. NC1775047