Learn More
enQuire Bioreagents Anti-GFP antibody | 50uL
Supplier:  enQuire Bioreagents AB29050UL
Encompass_Preferred
Rabbit polyclonal to GFP. Anti-GFP antibody (ab290) is a highly versatile antibody that gives a stronger signal than other anti-GFP antibodies available. On Western blot the antibody detects the GFP fraction from cell extracts expressing recombinant GFP fusion proteins and has also been shown to be useful on mouse sections fixed with formalin. In Immunocytochemistry, the antibody gives a very good signal on recombinant YES-GFP chimeras expressed in COS cells (McCabe et al. 1999 and figure below). It is routinely used in Immunoprecipitation (IP) and IP-Western protocols and has been used successfully in HRP Immunohistochemistry at 1:200 on whole-mount mouse embryos.
Catalog No. NC1666515