Learn More
ACROBiosystems ACROBiosystems Rabbit BCMA / TNFRSF17 Protein,His Tag
Supplier:  ACROBiosystems BCAR52H4100UG
Encompass_Preferred
Recombinant Protein;100UG;Rabbit BCMA, His Tag (BCA-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 1 - Asn 52 (Accession # G1SFN4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.9 kDa. The protein migrates as 12 kDa and 14 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human APRIL, Fc Tag (Cat. No. APL-H5267) at 5 μg/mL (100 μL/well) can bind Rabbit BCMA, His Tag (Cat. No. BCA-R52H4) with a linear range of 2-5 ng/mL (QC tested).;Immobilized Rabbit BCMA, His Tag (Cat. No. BCA-R52H4) at 2 μg/mL (100 μL/well) can bind Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) with a linear range of 0.4-6 ng/mL (Routinely tested).
Catalog No. 50-210-7271