Learn More
American Research Products Inc Rabbit Bone morphogenetic prot
Supplier:  American Research Products Inc CSBEL002736RB
Encompass
Rabbit Bone morphogenetic protein 2 (BMP2) ELISA kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27566