Learn More
Biomatik Corporation Rabbit Brain Derived Neurotrophic Factor (BDNF) ELISA Kit
Supplier:  Biomatik Corporation EKU02788
Encompass_Preferred
Rabbit Brain Derived Neurotrophic Factor (BDNF) ELISA Kit for detecting BDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G1SRK9. Target Synonyms: Neurotrophin; Abrineurin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4522