Learn More
American Research Products Inc Rabbit Endothelin 1 ET 1 ELIS
Supplier:  American Research Products Inc CSBE06951RB
Encompass
Rabbit Endothelin 1 ET-1 ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29098