Learn More
Rockland Immunochemicals Rabbit IgG (H&L) Antibody Pre-Adsorbed 1 mg
SDP
Supplier:  Rockland Immunochemicals 611201122
Encompass_Preferred
Anti-Rabbit IgG (H&L) generated in guinea pig detects rabbit Immunoglobulin G. Both the Heavy and Light chains of the antibody molecule are present. Representing approximately 75% of serum immunoglobulins IgG is the most abundant antibody isotype found in the circulation. IgG molecules are synthesized and secreted by plasma B cells. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3926