Learn More
American Research Products Inc Rabbit Immunoglobulin E IgE E
Supplier:  American Research Products Inc CSBE06948RB
Encompass
Rabbit Immunoglobulin E IgE ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26478