Learn More
American Research Products Inc Rabbit Immunoglobulin M IgM E
Supplier:  American Research Products Inc CSBE06950RB
Encompass
Rabbit Immunoglobulin M IgM ELISA Kit from CUSABIO Detection Range 15 6 ng/mL-4000 ng/mL Sensitivity 4 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26445