Learn More
American Research Products Inc Rabbit Interleukin 6 IL 6 ELI
Supplier:  American Research Products Inc CSBE06903RB
Encompass
Rabbit Interleukin 6 IL-6 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29634