Learn More
Cayman Chemical RabbIt ntI-Goat IgGSureLIg 1mg
SDP
Supplier:  Cayman Chemical 166981 MG
Encompass_Preferred
Rabbit anti-Goat IgG, secondary antibody, conjugated to SureLight P3Antibody: Rabbit polyclonal anti-Goat IgGIsotype: IgG Dye: SureLight P3 Excitation max. λ: 614 nm Emission max. λ: 662 nm Uses: Flow Cytometry, flurorescence-activated cell sorting, and protein microarrays Form: Conjugate supplied as a lyophilized powder.
Catalog No. 50-296-6715