Learn More
Cayman Chemical RabbIt ntIHA IgG DyLIght 5 1ea
SDP
Supplier:  Cayman Chemical 112581 EA
Encompass_Preferred
Rabbit anti-HA (Hemagglutinin) IgG:DyLight 550 Antibody: Rabbit polyclonal anti-HA (Hemagglutinin) (YPYDVPDYA) IgG Isotype: IgG Dye: DyLight 550 Excitation max. λ: 562 nm Emission max. λ: 576 nm Uses: Fluorescence-activated cell sorting, flow cytometry, and protein microarrays
Catalog No. 50-295-1113