Learn More
Cayman Chemical RabbIt ntIT7 IgG DyLIght 6
SDP
Supplier:  Cayman Chemical 16562100 UG
Encompass_Preferred
Rabbit anti-T7 IgG conjugated to DyLight 650 Antibody: Rabbit anti-T7-tag (MASMTGGQQMG) IgG Dye: DyLight 650 Excitation max. λ: 652 nm Emission max. λ: 672 nm Uses: Flow cytometry, fluorescence-activated cell sorting, microscopy, and TR-FRET
Catalog No. 50-296-8227